Anti-YIPF5 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26791-100EA 0 DKK
BIRBORB26791-100
Anti-YIPF5 Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:1000

Type: Primary
Antigen: YIPF5 (Yip1 domain family, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR