Anti-Muscarinic M3 Rec. Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26739-100EA 0 DKK
BIRBORB26739-100
Anti-Muscarinic M3 Rec. Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Muscarinic M3 Rec.
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR