Anti-Opioid Receptor, mu Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26717-100EA 0 DKK
BIRBORB26717-100
Anti-Opioid Receptor, mu Rabbit Polyclonal Antibody
Antistoffer
Immunochemistry: 1:2000; Western Blot: 1:500

Type: Primary
Antigen: Opioid Receptor, mu
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR