Anti-Fibrinogen Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26300-0.5EA 0 DKK
BIRBORB26300-0.5
Anti-Fibrinogen Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR