Anti-SERPINA7 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26169-1EA 0 DKK
BIRBORB26169-1
Anti-SERPINA7 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: SERPINA7 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR