Anti-CD8A Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26099-0.2EA 0 DKK
BIRBORB26099-0.2
Anti-CD8A Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR