Anti-PSA, Total Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26042-1EA 0 DKK
BIRBORB26042-1
Anti-PSA, Total Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
ELISA: 1:1000

Type: Primary
Antigen: PSA, Total
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR