Anti-CA125, Group B Mouse Monoclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26034-1EA 0 DKK
BIRBORB26034-1
Anti-CA125, Group B Mouse Monoclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CA125, Group B
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR