Anti-MUC1 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB26003-1EA 0 DKK
BIRBORB26003-1
Anti-MUC1 Mouse Monoclonal Antibody
Antistoffer
Immunochemistry: 50 ?g/ml

Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR