Anti-Glypican-1 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB25867-100EA 0 DKK
BIRBORB25867-100
Anti-Glypican-1 Mouse Monoclonal Antibody
Antistoffer
ELISA: 1-5 ?g/ml

Type: Primary
Antigen: Glypican-1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR