Anti-LTF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB25793-1.5EA 0 DKK
BIRBORB25793-1.5
Anti-LTF Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Immunochemistry: 1:500-1:5000; ELISA: 1:5000-1:100000; Western Blot: 1:10000-1:200000

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR