Anti-LTF Rabbit Polyclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB25790-1.5EA 0 DKK
BIRBORB25790-1.5
Anti-LTF Rabbit Polyclonal Antibody (Biotin)
Antistoffer
ELISA: 1:20000-1:400000; Western Blot: 1:500-1:5000; Immunofluorescence: 1:200-1:1000

Type: Primary
Antigen: LTF (lactotransferrin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR