Anti-APOC3 Goat Polyclonal Antibody (Biotin)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB25553-0.5EA 0 DKK
BIRBORB25553-0.5
Anti-APOC3 Goat Polyclonal Antibody (Biotin)
Antistoffer
ELISA: 1:200-1:20000
Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu