Anti-PLGF Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB25108-100EA 0 DKK
BIRBORB25108-100
Anti-PLGF Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PLGF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR