Anti-EGFR (Internal Domain) Mouse Monoclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB25008-0.25EA 0 DKK
BIRBORB25008-0.25
Anti-EGFR (Internal Domain) Mouse Monoclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: EGFR (Internal Domain)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR