Anti-Endothelin Mouse monoclonal antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB24788-100EA 0 DKK
BIRBORB24788-100
Anti-Endothelin Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: Endothelin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu