Anti-PSAT1 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24601-1EA 0 DKK
BIRBORB24601-1
Anti-PSAT1 Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSAT1 (phosphoserine aminotransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR