Anti-FSH (intact) Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24439-0.5EA 0 DKK
BIRBORB24439-0.5
Anti-FSH (intact) Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: FSH (intact)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR