Anti-INS Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB24430-1EA 0 DKK
BIRBORB24430-1
Anti-INS Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: INS (insulin)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu