Anti-AVP Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24314-100EA 0 DKK
BIRBORB24314-100
Anti-AVP Mouse Monoclonal Antibody
Antistoffer
ELISA: 1 ?g/ml

Type: Primary
Antigen: AVP (arginine vasopressin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR