Anti-Thymosin Alpha-1 Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24182-20EA 0 DKK
BIRBORB24182-20
Anti-Thymosin Alpha-1 Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:4000

Type: Primary
Antigen: Thymosin Alpha-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR