Anti-C-peptide Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24181-100EA 0 DKK
BIRBORB24181-100
Anti-C-peptide Rabbit Polyclonal Antibody
Antistoffer
RIA: 1:2000; Immunochemistry: 1:100; Western Blot: 1:100

Type: Primary
Antigen: C-peptide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR