Anti-HCV NS-4A Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24158-100EA 0 DKK
BIRBORB24158-100
Anti-HCV NS-4A Mouse monoclonal antibody
Antistoffer
Western Blot: 1:1000; Immunochemistry: 1:20

Type: Primary
Antigen: HCV NS-4A
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR