Anti-CMV Glycoprotein B Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB24141-100EA 0 DKK
BIRBORB24141-100
Anti-CMV Glycoprotein B Mouse Monoclonal Antibody
Antistoffer
  • Western Blot: 1:1000; ELISA: 1:10000

Type: Primary
Antigen: CMV Glycoprotein B
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR