Anti-Diphtheria Toxin Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB23989-1EA 0 DKK
BIRBORB23989-1
Anti-Diphtheria Toxin Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: Diphtheria Toxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR