Anti-Pneumocystis carinii Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB23589-100EA 0 DKK
BIRBORB23589-100
Anti-Pneumocystis carinii Mouse Monoclonal Antibody
Antistoffer
Immunofluorescence: 1:20
Type: Primary
Antigen: Pneumocystis carinii
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestil nu