Anti-Rabies Virus Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB23468-1EA 0 DKK
BIRBORB23468-1
Anti-Rabies Virus Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Rabies Virus
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR