Anti-Cholera Toxin Mouse Monoclonal Antibody
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB23320-1EA 0 DKK
BIRBORB23320-1
Anti-Cholera Toxin Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Cholera Toxin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu