Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB23208-1EA 0 DKK
BIRBORB23208-1
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu