Anti-N. meningitidis Rabbit Polyclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB23204-1EA 0 DKK
BIRBORB23204-1
Anti-N. meningitidis Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: N. meningitidis
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR