Anti-E. coli (all antigens) Goat Polyclonal Antibody (Biotin)

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB23084-1EA 0 DKK
BIRBORB23084-1
Anti-E. coli (all antigens) Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: E. coli (all antigens)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR