Anti-P. aeruginosa None Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB23053-1EA 0 DKK
BIRBORB23053-1
Anti-P. aeruginosa None Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: P. aeruginosa
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR