Anti-HA tag Mouse Monoclonal Antibody [clone: HA.C5]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22960-100EA 0 DKK
BIRBORB22960-100
Anti-HA tag Mouse Monoclonal Antibody [clone: HA.C5]
Antistoffer
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: HA.C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR