Anti-WNT3A Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22925-5EA 0 DKK
BIRBORB22925-5 BIRBORB22925-20 BIRBORB22925-100
Anti-WNT3A Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: WNT3A (wingless-type MMTV integration site family, member 3A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR