Anti-KIR2DL1 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22851-5EA 0 DKK
BIRBORB22851-5 BIRBORB22851-20 BIRBORB22851-100
Anti-KIR2DL1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: KIR2DL1 (killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR