Anti-ISG15 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22834-20EA 0 DKK
BIRBORB22834-20 BIRBORB22834-5 BIRBORB22834-100
Anti-ISG15 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: ISG15 (ISG15 ubiquitin-like modifier)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR