Anti-EPHA2 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22820-100EA 0 DKK
BIRBORB22820-100 BIRBORB22820-20 BIRBORB22820-5
Anti-EPHA2 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: EPHA2 (EPH receptor A2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR