Anti-CSTB Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22818-20EA 0 DKK
BIRBORB22818-20 BIRBORB22818-100 BIRBORB22818-5
Anti-CSTB Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: CSTB (cystatin B (stefin B))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR