Anti-CFL1 Mouse monoclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22812-20EA 0 DKK
BIRBORB22812-20 BIRBORB22812-5 BIRBORB22812-100
Anti-CFL1 Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR