Anti-Influenza H1N1 Mouse Monoclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22720-10EA 0 DKK
BIRBORB22720-10 BIRBORB22720-50
Anti-Influenza H1N1 Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: Influenza H1N1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Virus
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR