Anti-CD4 Mouse Monoclonal Antibody (Biotin)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB22643-1EA 0 DKK
BIRBORB22643-1 BIRBORB22643-500
Anti-CD4 Mouse Monoclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human