Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein))

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22637-1EA 0 DKK
BIRBORB22637-1 BIRBORB22637-500
Anti-CD2 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR