Anti-CSF3 Mouse Monoclonal Antibody (FITC (Fluorescein))
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BIRBORB22590-1EA 0 DKK
BIRBORB22590-1 BIRBORB22590-500
Anti-CSF3 Mouse Monoclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: CSF3 (colony stimulating factor 3 (granulocyte))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human