Anti-MOB4 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22568-100EA 0 DKK
BIRBORB22568-100
Anti-MOB4 Goat Polyclonal Antibody
Antistoffer
ELISA: 1:4000; Western Blot: 0.3-1 ?g/ml; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: MOB4 (MOB family member 4, phocein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR