Anti-ITGA11 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22549-100EA 0 DKK
BIRBORB22549-100
Anti-ITGA11 Goat Polyclonal Antibody
Antistoffer
ELISA: 1:16000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: ITGA11 (integrin, alpha 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR