Anti-ZEB2 Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22539-100EA 0 DKK
BIRBORB22539-100
Anti-ZEB2 Goat Polyclonal Antibody
Antistoffer
ELISA: 1:32000

Type: Primary
Antigen: ZEB2 (zinc finger E-box binding homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR