Anti-IDH3G Goat Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB22492-100EA 0 DKK
BIRBORB22492-100
Anti-IDH3G Goat Polyclonal Antibody
Antistoffer
ELISA: 1:16000; Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: IDH3G (isocitrate dehydrogenase 3 (NAD+) gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR