Anti-Complement C4 beta chain Rabbit Polyclonal Antibody

Leverandør: Biorbyt
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB13138-100EA 2550 DKK
BIRBORB13138-100
Anti-Complement C4 beta chain Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:1000

Type: Primary
Antigen: Complement C4 beta chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR