Anti-CST3 Rabbit polyclonal antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB13133-100EA 0 DKK
BIRBORB13133-100
Anti-CST3 Rabbit polyclonal antibody
Antistoffer
Western Blot: 1:1000

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR