Anti-LTA Rabbit Polyclonal Antibody

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BIRBORB13129-100EA 0 DKK
BIRBORB13129-100
Anti-LTA Rabbit Polyclonal Antibody
Antistoffer
Western Blot: 1:1000; Immunofluorescence: 1:250

Type: Primary
Antigen: LTA (lymphotoxin alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR